Assay Details
| Gene Symbol: LOC4346592 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015755934.2 | XP_015611420.1 | 1 | 133 | 88 | |
| GenBank mRNA | AK070953.1 | - | 1 | 59 | 88 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015755934.2 transcripts.
Set Membership: |
|

LOC4346592
Genomic Map