Assay Details
| Gene Symbol: LOC4346740 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756394.1 | XP_015611880.1 | 1 - 2 | 2560 | 74 | |
| GenBank mRNA | AK120843.1 | - | 1 - 2 | 2568 | 74 | |
More Information
Set Membership: |
|

LOC4346740
Genomic Map