Assay Details
| Gene Symbol: LOC4346874 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756834.2 | XP_015612320.1 | 1 - 2 | 161 | 77 | |
| GenBank mRNA | AK063494.1 | - | 1 - 2 | 59 | 77 | |
| AK111465.1 | - | 1 - 2 | 56 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015756834.2 transcripts.
Set Membership: |
|

LOC4346874
Genomic Map