Assay Details
| Gene Symbol: LOC4347303 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795950.2 | XP_015651436.1 | 2 | 1525 | 109 | |
| XM_015795951.2 | XP_015651437.1 | 2 | 1518 | 109 | ||
| GenBank mRNA | AK060708.1 | - | 1 | 1551 | 109 | |
More Information
Set Membership: |
|

LOC4347303
Genomic Map