Assay Details
| Gene Symbol: LOC4347310 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756311.2 | XP_015611797.1 | 1 - 2 | 418 | 136 | |
| XM_015756312.2 | XP_015611798.1 | 1 - 2 | 430 | 136 | ||
| GenBank mRNA | AK109671.1 | - | 1 - 2 | 430 | 136 | |
More Information
Set Membership: |
|

LOC4347310
Genomic Map