Assay Details
| Gene Symbol: LOC4347320 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756225.2 | XP_015611711.1 | 1 - 2 | 906 | 60 | |
| GenBank mRNA | AK064135.1 | - | 1 - 2 | 900 | 60 | |
More Information
Set Membership: |
|

LOC4347320
Genomic Map