Assay Details
| Gene Symbol: LOC4347572 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756480.2 | XP_015611966.1 | 10 - 11 | 3424 | 134 | |
| GenBank mRNA | AK099978.1 | - | 10 - 11 | 3405 | 134 | |
More Information
Set Membership: |
|

LOC4347572
Genomic Map