Assay Details
| Gene Symbol: LOC4347638 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015755237.2 | XP_015610723.1 | 3 - 4 | 588 | 58 | |
| GenBank mRNA | AB097460.1 | - | 3 - 4 | 460 | 58 | |
| AK073610.1 | - | 3 - 4 | 532 | 58 | ||
| AK104977.1 | - | 3 - 4 | 476 | 58 | ||
More Information
Set Membership: |
|

LOC4347638
Genomic Map