Assay Details
| Gene Symbol: LOC4347638 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015755237.2 | XP_015610723.1 | 4 - 5 | 697 | 60 | |
| GenBank mRNA | AB097460.1 | - | 4 - 5 | 569 | 60 | |
| AK073610.1 | - | 4 - 5 | 641 | 60 | ||
| AK104977.1 | - | 4 - 5 | 585 | 60 | ||
More Information
Set Membership: |
|

LOC4347638
Genomic Map