Assay Details
| Gene Symbol: LOC4347778 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015755334.1 | XP_015610820.1 | 1 - 2 | 122 | 72 | |
| GenBank mRNA | AK103727.1 | - | 1 - 2 | 112 | 72 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015755334.1 transcripts.
Set Membership: |
|

LOC4347778
Genomic Map