Assay Details
| Gene Symbol: LOC4347778 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015755333.2 | XP_015610819.1 | 2 - 3 | 1211 | 64 | |
| XM_015755334.1 | XP_015610820.1 | 2 - 3 | 1228 | 64 | ||
| GenBank mRNA | AK103727.1 | - | 2 - 3 | 1218 | 64 | |
More Information
Set Membership: |
|

LOC4347778
Genomic Map