Assay Details
| Gene Symbol: LOC4347778 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015755333.2 | XP_015610819.1 | 3 - 4 | 1288 | 100 | |
| XM_015755334.1 | XP_015610820.1 | 3 - 4 | 1305 | 100 | ||
| GenBank mRNA | AK103727.1 | - | 3 - 4 | 1295 | 100 | |
More Information
Set Membership: |
|

LOC4347778
Genomic Map