Assay Details
| Gene Symbol: LOC4347787 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756522.2 | XP_015612008.1 | 13 - 14 | 1530 | 102 | |
| GenBank mRNA | AK066732.1 | - | 13 - 14 | 1537 | 102 | |
| AK073242.1 | - | 13 - 14 | 1536 | 102 | ||
More Information
Set Membership: |
|

LOC4347787
Genomic Map