Assay Details
| Gene Symbol: LOC4348618 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757689.2 | XP_015613175.1 | 5 - 6 | 663 | 66 | |
| GenBank mRNA | AK067754.1 | - | 5 - 6 | 627 | 66 | |
More Information
Set Membership: |
|

LOC4348618
Genomic Map