Assay Details
| Gene Symbol: LOC4348618 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757689.2 | XP_015613175.1 | 6 - 7 | 810 | 92 | |
| GenBank mRNA | AK067754.1 | - | 6 - 7 | 774 | 92 | |
More Information
Set Membership: |
|

LOC4348618
Genomic Map