Assay Details
| Gene Symbol: LOC4348723 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759390.2 | XP_015614876.1 | 6 - 7 | 780 | 106 | |
| GenBank mRNA | AK240989.1 | - | 2023 | 106 | ||
| AK287809.1 | - | 2034 | 106 | |||
| AY224551.1 | - | 6 - 7 | 652 | 106 | ||
More Information
Set Membership: |
|

LOC4348723
Genomic Map