Assay Details
| Gene Symbol: LOC4348901 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757764.2 | XP_015613250.1 | 2 - 3 | 712 | 73 | |
| GenBank mRNA | AK119558.1 | - | 2 - 3 | 591 | 73 | |
| GQ229485.1 | - | 2 - 3 | 434 | 73 | ||
More Information
Set Membership: |
|

LOC4348901
Genomic Map