Assay Details
| Gene Symbol: LOC4348965 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758987.1 | XP_015614473.1 | 14 - 15 | 2545 | 75 | |
| XM_015758988.2 | XP_015614474.1 | 14 - 15 | 2545 | 75 | ||
| GenBank mRNA | AK120278.1 | - | 14 - 15 | 2557 | 75 | |
More Information
Set Membership: |
|

LOC4348965
Genomic Map