Assay Details
| Gene Symbol: LOC4348965 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758987.1 | XP_015614473.1 | 17 - 18 | 2770 | 105 | |
| GenBank mRNA | AK120278.1 | - | 17 - 18 | 2782 | 105 | |
More Information
Set Membership: |
|

LOC4348965
Genomic Map