Assay Details
| Gene Symbol: LOC4349025 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759374.2 | XP_015614860.1 | 2 | 1439 | 125 | |
| GenBank mRNA | AK061011.1 | - | 1 | 948 | 125 | |
More Information
Set Membership: |
|

LOC4349025
Genomic Map