Assay Details
| Gene Symbol: LOC4349516 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758832.1 | XP_015614318.1 | 2 - 3 | 286 | 71 | |
| GenBank mRNA | AK070527.1 | - | 2 - 3 | 224 | 71 | |
| AK099219.1 | - | 2 - 3 | 224 | 71 | ||
| AK099267.1 | - | 2 - 3 | 206 | 71 | ||
| AK105726.1 | - | 2 - 3 | 216 | 71 | ||
More Information
Set Membership: |
|

LOC4349516
Genomic Map