Assay Details
| Gene Symbol: LOC4349540 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759648.2 | XP_015615134.1 | 1 - 2 | 896 | 80 | |
| GenBank mRNA | AK101720.1 | - | 1 - 2 | 895 | 80 | |
More Information
Set Membership: |
|

LOC4349540
Genomic Map