Assay Details
| Gene Symbol: LOC4349978 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761494.2 | XP_015616980.1 | 12 - 13 | 2308 | 96 | |
| XM_015761495.2 | XP_015616981.1 | 12 - 13 | 2325 | 96 | ||
| GenBank mRNA | AK060444.1 | - | 1 - 2 | 115 | 96 | |
| AK070280.1 | - | 12 - 13 | 2331 | 96 | ||
| AK101281.1 | - | 11 - 12 | 2456 | 96 | ||
More Information
Set Membership: |
|

LOC4349978
Genomic Map