Assay Details
| Gene Symbol: LOC4349978 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761495.2 | XP_015616981.1 | 13 - 14 | 2509 | 122 | |
| GenBank mRNA | AK060444.1 | - | 2 - 3 | 299 | 122 | |
| AK070280.1 | - | 13 - 14 | 2515 | 122 | ||
| AK101281.1 | - | 12 - 13 | 2640 | 122 | ||
More Information
Set Membership: |
|

LOC4349978
Genomic Map