Assay Details
| Gene Symbol: LOC4349978 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761494.2 | XP_015616980.1 | 3 - 4 | 476 | 76 | |
| XM_015761495.2 | XP_015616981.1 | 3 - 4 | 493 | 76 | ||
| GenBank mRNA | AK070280.1 | - | 3 - 4 | 499 | 76 | |
| AK101281.1 | - | 3 - 4 | 501 | 76 | ||
More Information
Set Membership: |
|

LOC4349978
Genomic Map