Assay Details
| Gene Symbol: LOC4349978 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761494.2 | XP_015616980.1 | 10 - 11 | 1848 | 120 | |
| XM_015761495.2 | XP_015616981.1 | 10 - 11 | 1865 | 120 | ||
| GenBank mRNA | AK070280.1 | - | 10 - 11 | 1871 | 120 | |
| AK101281.1 | - | 9 - 10 | 1996 | 120 | ||
More Information
Set Membership: |
|

LOC4349978
Genomic Map