Assay Details
| Gene Symbol: LOC4350002 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761828.1 | XP_015617314.1 | 5 - 6 | 895 | 83 | |
| GenBank mRNA | AB373950.1 | - | 5 - 6 | 881 | 83 | |
| AK059538.1 | - | 3 - 4 | 544 | 83 | ||
| AK068268.1 | - | 5 - 6 | 883 | 83 | ||
| AY987391.1 | - | 5 - 6 | 883 | 83 | ||
More Information
Set Membership: |
|

LOC4350002
Genomic Map