Assay Details
| Gene Symbol: LOC4350002 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761828.1 | XP_015617314.1 | 9 - 10 | 1389 | 79 | |
| GenBank mRNA | AB373950.1 | - | 9 - 10 | 1375 | 79 | |
| AK059538.1 | - | 7 - 8 | 1038 | 79 | ||
| AK068268.1 | - | 9 - 10 | 1377 | 79 | ||
More Information
Set Membership: |
|

LOC4350002
Genomic Map