Assay Details
| Gene Symbol: LOC4328224 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770971.2 | XP_015626457.1 | 9 - 10 | 1596 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4328224| Gene Symbol: LOC4328224 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770971.2 | XP_015626457.1 | 9 - 10 | 1596 | 64 | |
Set Membership: |
|