Assay Details
| Gene Symbol: LOC4328488 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770639.2 | XP_015626125.1 | 6 - 7 | 829 | 68 | |
| GenBank mRNA | AK060768.1 | - | 1 - 2 | 102 | 68 | |
| AK066421.1 | - | 1 - 2 | 42 | 68 | ||
More Information
Set Membership: |
|

LOC4328488
Genomic Map