Assay Details
| Gene Symbol: LOC4328488 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770639.2 | XP_015626125.1 | 10 - 11 | 1244 | 148 | |
| GenBank mRNA | AK060768.1 | - | 5 - 6 | 517 | 148 | |
| AK066421.1 | - | 5 - 6 | 457 | 148 | ||
More Information
Set Membership: |
|

LOC4328488
Genomic Map