Assay Details
| Gene Symbol: LOC4328552 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770922.2 | XP_015626408.1 | 1 - 2 | 260 | 112 | |
| GenBank mRNA | AK071794.1 | - | 1 - 2 | 222 | 112 | |
| AK105011.2 | - | 1 - 2 | 221 | 112 | ||
More Information
Set Membership: |
|

LOC4328552
Genomic Map