Assay Details
| Gene Symbol: LOC4328552 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770922.2 | XP_015626408.1 | 2 - 3 | 378 | 108 | |
| GenBank mRNA | AK059804.1 | - | 1 - 2 | 83 | 108 | |
| AK071794.1 | - | 2 - 3 | 340 | 108 | ||
| AK105011.2 | - | 2 - 3 | 339 | 108 | ||
More Information
Set Membership: |
|

LOC4328552
Genomic Map