Assay Details
| Gene Symbol: LOC4328552 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770922.2 | XP_015626408.1 | 3 - 4 | 607 | 117 | |
| GenBank mRNA | AK059804.1 | - | 2 - 3 | 312 | 117 | |
| AK071794.1 | - | 3 - 4 | 569 | 117 | ||
| AK105011.2 | - | 3 - 4 | 568 | 117 | ||
| AK105739.1 | - | 714 | 117 | |||
More Information
Set Membership: |
|

LOC4328552
Genomic Map