Assay Details
| Gene Symbol: LOC4328628 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768249.2 | XP_015623735.1 | 1 - 2 | 192 | 83 | |
| GenBank mRNA | AK058544.1 | - | 1 - 2 | 154 | 83 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015768249.2 transcripts.
Set Membership: |
|

LOC4328628
Genomic Map