Assay Details
| Gene Symbol: LOC4328628 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768248.2 | XP_015623734.1 | 2 - 3 | 864 | 152 | |
| XM_015768249.2 | XP_015623735.1 | 2 - 3 | 430 | 152 | ||
| GenBank mRNA | AK058544.1 | - | 2 - 3 | 392 | 152 | |
More Information
Set Membership: |
|

LOC4328628
Genomic Map