Assay Details
| Gene Symbol: LOC4328656 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768436.2 | XP_015623922.1 | 1 - 2 | 110 | 158 | |
| GenBank mRNA | AK107368.1 | - | 1 - 2 | 70 | 157 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015768436.2 transcripts.
Set Membership: |
|

LOC4328656
Genomic Map