Assay Details
| Gene Symbol: LOC4328656 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768434.2 | XP_015623920.1 | 2 - 3 | 734 | 77 | |
| XM_015768435.2 | XP_015623921.1 | 2 - 3 | 735 | 77 | ||
| XM_015768436.2 | XP_015623922.1 | 2 - 3 | 738 | 77 | ||
| GenBank mRNA | AK107368.1 | - | 2 - 3 | 697 | 77 | |
More Information
Set Membership: |
|

LOC4328656
Genomic Map