Assay Details
| Gene Symbol: LOC4328986 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767452.2 | XP_015622938.1 | 1 - 2 | 701 | 136 | |
| GenBank mRNA | AK072271.1 | - | 1 - 2 | 669 | 136 | |
More Information
Set Membership: |
|

LOC4328986
Genomic Map