Assay Details
| Gene Symbol: LOC4329076 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771222.2 | XP_015626708.1 | 5 | 3310 | 146 | |
| GenBank mRNA | AK069078.1 | - | 1 | 2643 | 146 | |
More Information
Set Membership: |
|

LOC4329076
Genomic Map