Assay Details
| Gene Symbol: LOC4329333 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772156.2 | XP_015627642.1 | 1 - 2 | 443 | 67 | |
| XM_015772157.2 | XP_015627643.1 | 1 - 2 | 438 | 67 | ||
| GenBank mRNA | AK120593.1 | - | 1 - 2 | 340 | 67 | |
More Information
Set Membership: |
|

LOC4329333
Genomic Map