Assay Details
| Gene Symbol: LOC4329333 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772156.2 | XP_015627642.1 | 5 - 6 | 1135 | 127 | |
| XM_015772157.2 | XP_015627643.1 | 5 - 6 | 1112 | 127 | ||
| GenBank mRNA | AK120593.1 | - | 5 - 6 | 1014 | 127 | |
More Information
Set Membership: |
|

LOC4329333
Genomic Map