Assay Details
| Gene Symbol: LOC4329519 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768155.2 | XP_015623641.1 | 11 - 12 | 1324 | 63 | |
| XM_015768156.2 | XP_015623642.1 | 10 - 11 | 1197 | 63 | ||
| GenBank mRNA | AK058905.1 | - | 3 - 4 | 223 | 63 | |
| AK071231.1 | - | 10 - 11 | 1157 | 63 | ||
| AK288370.1 | - | 11 - 12 | 1325 | 63 | ||
| BT130382.1 | - | 2 - 3 | 117 | 63 | ||
More Information
Set Membership: |
|

LOC4329519
Genomic Map