Assay Details
| Gene Symbol: LOC4329519 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768156.2 | XP_015623642.1 | 1 - 2 | 324 | 98 | |
| GenBank mRNA | AK071231.1 | - | 1 - 2 | 284 | 98 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015768156.2 transcripts.
Set Membership: |
|

LOC4329519
Genomic Map