Assay Details
| Gene Symbol: LOC4329519 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768155.2 | XP_015623641.1 | 6 - 7 | 864 | 130 | |
| XM_015768156.2 | XP_015623642.1 | 5 - 6 | 737 | 130 | ||
| GenBank mRNA | AK071231.1 | - | 5 - 6 | 697 | 130 | |
| AK288370.1 | - | 6 - 7 | 865 | 130 | ||
More Information
Set Membership: |
|

LOC4329519
Genomic Map