Assay Details
| Gene Symbol: LOC4329529 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770319.2 | XP_015625805.1 | 1 | 1459 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4329529| Gene Symbol: LOC4329529 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770319.2 | XP_015625805.1 | 1 | 1459 | 72 | |
Set Membership: |
|