Assay Details
| Gene Symbol: LOC4330182 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771452.1 | XP_015626938.1 | 1 | 1935 | 123 | |
| GenBank mRNA | AK064950.1 | - | 1 | 1753 | 123 | |
| AK098925.1 | - | 1 | 1754 | 123 | ||
| AK104793.1 | - | 1 | 1753 | 123 | ||
| AK106154.1 | - | 1 | 1753 | 123 | ||
More Information
Set Membership: |
|

LOC4330182
Genomic Map