Assay Details
| Gene Symbol: LOC4330184 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767746.2 | XP_015623232.1 | 3 - 4 | 465 | 106 | |
| GenBank mRNA | AK101814.1 | - | 3 - 4 | 458 | 106 | |
More Information
Set Membership: |
|

LOC4330184
Genomic Map