Assay Details
| Gene Symbol: LOC4330292 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772114.2 | XP_015627600.1 | 2 - 3 | 618 | 85 | |
| GenBank mRNA | AK242994.1 | - | 2 - 3 | 578 | 85 | |
| BT130241.1 | - | 1 - 2 | 118 | 85 | ||
More Information
Set Membership: |
|

LOC4330292
Genomic Map