Assay Details
| Gene Symbol: LOC4330310 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767520.2 | XP_015623006.1 | 4 - 5 | 675 | 89 | |
| XM_015767521.2 | XP_015623007.1 | 4 - 5 | 669 | 89 | ||
| XM_015767522.2 | XP_015623008.1 | 3 - 4 | 387 | 89 | ||
| XM_015767523.2 | XP_015623009.1 | 4 - 5 | 675 | 89 | ||
| GenBank mRNA | AK241359.1 | - | 4 - 5 | 675 | 89 | |
More Information
Set Membership: |
|

LOC4330310
Genomic Map